Tisk…

Anti-IL4 Mouse monoclonal antibody [clone: 4D9]

ABCAAB13779-100EA 15370 CZK
ABCAAB13779-100
Anti-IL4 Mouse monoclonal antibody [clone: 4D9]
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: monoclonal
Clone: 4D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR