Tisk…

Anti-STAM Mouse monoclonal antibody [clone: 29C678]

ABCAAB13797-100EA 15370 CZK
ABCAAB13797-100
Anti-STAM Mouse monoclonal antibody [clone: 29C678]
Type: Primary
Antigen: STAM (signal transducing adaptor molecule (SH3 domain and ITAM motif) 1)
Clonality: monoclonal
Clone: 29C678
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR