Tisk…

Anti-Hepatitis C Virus antigen Mouse Monoclonal Antibody [clone: H25]

Dodavatel: Abcam
ABCAAB13832-100EA 21300 CZK
ABCAAB13832-100
Anti-Hepatitis C Virus antigen Mouse Monoclonal Antibody [clone: H25]
Protilátky
Type: Primary
Antigen: Hepatitis C Virus antigen
Clonality: monoclonal
Clone: H25
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR