Tisk…
Anti-TRDMT1 Mouse monoclonal antibody [clone: 102B1259.2]
ABCAAB13899-100EA 16080 CZK
ABCAAB13899-100
Anti-TRDMT1 Mouse monoclonal antibody [clone: 102B1259.2]
Type: Primary
Antigen: TRDMT1 (tRNA aspartic acid methyltransferase 1)
Clonality: monoclonal
Clone: 102B1259.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse