Tisk…

Anti-NIFK Rabbit polyclonal antibody

ABCAAB13912-200EA 15370 CZK
ABCAAB13912-200
Anti-NIFK Rabbit polyclonal antibody
Type: Primary
Antigen: NIFK (nucleolar protein interacting with the FHA domain of MKI67)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR