Tisk…

Anti-SOCS4 Rabbit polyclonal antibody

ABCAAB13948-100EA 15770 CZK
ABCAAB13948-100
Anti-SOCS4 Rabbit polyclonal antibody
Type: Primary
Antigen: SOCS4 (suppressor of cytokine signaling 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR