Tisk…

Anti-SOCS6 Rabbit polyclonal antibody

ABCAAB13950-100EA 15770 CZK
ABCAAB13950-100
Anti-SOCS6 Rabbit polyclonal antibody
Type: Primary
Antigen: SOCS6 (suppressor of cytokine signaling 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR