Tisk…

Anti-SIGLEC7 Rabbit polyclonal antibody

ABCAAB13964-100EA 0 CZK
ABCAAB13964-100
Anti-SIGLEC7 Rabbit polyclonal antibody
Type: Primary
Antigen: SIGLEC7 (sialic acid binding Ig-like lectin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR