Tisk…

Anti-PTN Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB14025-50EA 21330 CZK
ABCAAB14025-50
Anti-PTN Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PTN (pleiotrophin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR