Tisk…

Anti-IL2 Chicken Polyclonal Antibody

Dodavatel: Abcam
ABCAAB14036-50EA 22550 CZK
ABCAAB14036-50
Anti-IL2 Chicken Polyclonal Antibody
Protilátky
Type: Primary
Antigen: IL2 (interleukin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR