Tisk…

Anti-CFL2 Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB14134-50EA 21140 CZK
ABCAAB14134-50
Anti-CFL2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CFL2 (cofilin 2 (muscle))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR