Tisk…

Anti-GCG Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB14183-25EA 22340 CZK
ABCAAB14183-25
Anti-GCG Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: GCG (glucagon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR