Tisk…

Anti-Prostatic Acid Phosphatase Goat polyclonal antibody

ABCAAB14192-100EA 16280 CZK
ABCAAB14192-100
Anti-Prostatic Acid Phosphatase Goat polyclonal antibody
Type: Primary
Antigen: Prostatic Acid Phosphatase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR