Tisk…

Anti-Prion protein PrP Mouse Monoclonal Antibody [clone: 7D9]

Dodavatel: Abcam
ABCAAB14219-100EA 21640 CZK
ABCAAB14219-100
Anti-Prion protein PrP Mouse Monoclonal Antibody [clone: 7D9]
Protilátky
Type: Primary
Antigen: Prion protein PrP
Clonality: monoclonal
Clone: 7D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR