Tisk…

Anti-APOA1 Chicken polyclonal antibody

ABCAAB14229-50EA 17670 CZK
ABCAAB14229-50
Anti-APOA1 Chicken polyclonal antibody
Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR