Tisk…

Anti-CDK4 Chicken polyclonal antibody

ABCAAB14242-50EA 16480 CZK
ABCAAB14242-50
Anti-CDK4 Chicken polyclonal antibody
Type: Primary
Antigen: CDK4 (cyclin-dependent kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR