Tisk…

Anti-EIF4EBP2 Chicken polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB14295-50EA 24390 CZK
ABCAAB14295-50
Anti-EIF4EBP2 Chicken polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: EIF4EBP2 (eukaryotic translation initiation factor 4E binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR