Tisk…

Anti-Natriuretic Peptide Receptor B Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB14357-50EA 21330 CZK
ABCAAB14357-50
Anti-Natriuretic Peptide Receptor B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Natriuretic Peptide Receptor B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR