Tisk…

Anti-DEFB105A Rabbit polyclonal antibody

ABCAAB14363-50EA 16080 CZK
ABCAAB14363-50
Anti-DEFB105A Rabbit polyclonal antibody
Type: Primary
Antigen: DEFB105A (defensin, beta 105A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR