Tisk…

Anti-CTBP1 Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB14411-50EA 21140 CZK
ABCAAB14411-50
Anti-CTBP1 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: CTBP1 (C-terminal binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR