Tisk…

Anti-pro Guanylin Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB14427-50EA 21360 CZK
ABCAAB14427-50
Anti-pro Guanylin Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: pro Guanylin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR