Tisk…

Anti-Guanylate Cyclase Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB14438-50EA 21140 CZK
ABCAAB14438-50
Anti-Guanylate Cyclase Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Guanylate Cyclase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR