Tisk…

Anti-NPPA Mouse Monoclonal Antibody [clone: L11-11-D6-B4]

Dodavatel: Abcam
ABCAAB14442-50EA 28070 CZK
ABCAAB14442-50
Anti-NPPA Mouse Monoclonal Antibody [clone: L11-11-D6-B4]
Protilátky
Type: Primary
Antigen: NPPA (natriuretic peptide A)
Clonality: monoclonal
Clone: L11-11-D6-B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR