Tisk…

Anti-GCDFP 15 Mouse monoclonal antibody unconjugated [clone: 0.N.307]

Dodavatel: Abcam
ABCAAB14478-250EA 26140 CZK
ABCAAB14478-250
Anti-GCDFP 15 Mouse monoclonal antibody unconjugated [clone: 0.N.307]
Protilátky
Type: Primary
Antigen: GCDFP 15
Clonality: monoclonal
Clone: 0.N.307
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR