Tisk…

Anti-Macrophage inflammatory protein 5 Rabbit polyclonal antibody

ABCAAB14532-50EA 16080 CZK
ABCAAB14532-50
Anti-Macrophage inflammatory protein 5 Rabbit polyclonal antibody
Type: Primary
Antigen: Macrophage inflammatory protein 5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR