Tisk…

Anti-Mitotic proteins Mouse Monoclonal Antibody [clone: 0.T.181]

Dodavatel: Abcam
ABCAAB14581-50EA 23080 CZK
ABCAAB14581-50
Anti-Mitotic proteins Mouse Monoclonal Antibody [clone: 0.T.181]
Protilátky
Type: Primary
Antigen: Mitotic proteins
Clonality: monoclonal
Clone: 0.T.181
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR