Tisk…

Anti-DICER1 Mouse monoclonal antibody unconjugated [clone: 13D6]

Dodavatel: Abcam
ABCAAB14601-100EA 26140 CZK
ABCAAB14601-100
Anti-DICER1 Mouse monoclonal antibody unconjugated [clone: 13D6]
Protilátky
Type: Primary
Antigen: DICER1 (dicer 1, ribonuclease type III)
Clonality: monoclonal
Clone: 13D6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR