Tisk…

Anti-VDAC1 Mouse Monoclonal Antibody [clone: 20B12AF2]

Dodavatel: Abcam
ABCAAB14734-100EA 24480 CZK
ABCAAB14734-100
Anti-VDAC1 Mouse Monoclonal Antibody [clone: 20B12AF2]
Protilátky
Type: Primary
Antigen: VDAC1 (voltage-dependent anion channel 1)
Clonality: monoclonal
Clone: 20B12AF2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR