Tisk…

Anti-FAF2 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB14759-100EA 20130 CZK
ABCAAB14759-100
Anti-FAF2 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FAF2 (Fas associated factor family member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR