Tisk…

Anti-KIR2DL3 Mouse monoclonal antibody [clone: 190IIC311]

ABCAAB14825-100EA 16080 CZK
ABCAAB14825-100
Anti-KIR2DL3 Mouse monoclonal antibody [clone: 190IIC311]
Type: Primary
Antigen: KIR2DL3 (killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3)
Clonality: monoclonal
Clone: 190IIC311
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR