Tisk…

Anti-BEST1 Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB14928-50EA 22520 CZK
ABCAAB14928-50
Anti-BEST1 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: BEST1 (bestrophin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR