Tisk…

Anti-CDKN2A Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB14932-100EA 21140 CZK
ABCAAB14932-100
Anti-CDKN2A Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR