Tisk…
Anti-NCL Mouse Monoclonal Antibody [clone: 4i51]
Dodavatel: Abcam
ABCAAB14966-25EA 27760 CZK
ABCAAB14966-25
Anti-NCL Mouse Monoclonal Antibody [clone: 4i51]
Protilátky
Type: Primary
Antigen: NCL (nucleolin)
Clonality: monoclonal
Clone: 4i51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat