Tisk…

Anti-PKP1 Mouse monoclonal antibody [clone: 3G250]

ABCAAB15006-50EA 16080 CZK
ABCAAB15006-50
Anti-PKP1 Mouse monoclonal antibody [clone: 3G250]
Type: Primary
Antigen: PKP1 (plakophilin 1 (ectodermal dysplasia/skin fragility syndrome))
Clonality: monoclonal
Clone: 3G250
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR