Tisk…

Anti-CLDN1 Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB15098-500EA 23870 CZK
ABCAAB15098-500
Anti-CLDN1 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: CLDN1 (claudin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR