Tisk…

Anti-AQP4 Rabbit polyclonal antibody

ABCAAB15118-25EA 17160 CZK
ABCAAB15118-25
Anti-AQP4 Rabbit polyclonal antibody
Type: Primary
Antigen: AQP4 (aquaporin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR