Tisk…

Anti-AQP7 Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB15122-25EA 21360 CZK
ABCAAB15122-25
Anti-AQP7 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: AQP7 (aquaporin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR