Tisk…

Anti-TXNRD2 Rabbit polyclonal antibody

ABCAAB15174-7EA 16280 CZK
ABCAAB15174-7
Anti-TXNRD2 Rabbit polyclonal antibody
Type: Primary
Antigen: TXNRD2 (thioredoxin reductase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR