Tisk…

Anti-APAF1 Rabbit polyclonal antibody

ABCAAB15178-7EA 17330 CZK
ABCAAB15178-7
Anti-APAF1 Rabbit polyclonal antibody
Type: Primary
Antigen: APAF1 (apoptotic peptidase activating factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR