Tisk…

Anti-GJA1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB15189-500EA 21330 CZK
ABCAAB15189-500
Anti-GJA1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GJA1 (gap junction protein, alpha 1, 43kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR