Tisk…

Anti-NOS1 Rabbit polyclonal antibody

ABCAAB15204-7EA 16930 CZK
ABCAAB15204-7
Anti-NOS1 Rabbit polyclonal antibody
Type: Primary
Antigen: NOS1 (nitric oxide synthase 1 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR