Tisk…

Anti-Cytokeratin 8+18 Mouse monoclonal antibody [clone: SP141]

ABCAAB15224-1EA 15650 CZK
ABCAAB15224-1
Anti-Cytokeratin 8+18 Mouse monoclonal antibody [clone: SP141]
Type: Primary
Antigen: Cytokeratin 8+18
Clonality: monoclonal
Clone: SP141
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR