Tisk…

Anti-WT1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB15249-500EA 22060 CZK
ABCAAB15249-500
Anti-WT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: WT1 (Wilms tumor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR