Tisk…

Anti-ACTB Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB15263-1EA 22310 CZK
ABCAAB15263-1
Anti-ACTB Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR