Tisk…

Anti-DMD Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB15277-500EA 23960 CZK
ABCAAB15277-500
Anti-DMD Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DMD (dystrophin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR