Tisk…

Anti-GRIK2 + GRIK3 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB15307-500EA 21850 CZK
ABCAAB15307-500
Anti-GRIK2 + GRIK3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GRIK2 + GRIK3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR