Tisk…

Anti-MelanA Rabbit polyclonal antibody

ABCAAB15468-500EA 0 CZK
ABCAAB15468-500
Anti-MelanA Rabbit polyclonal antibody
Type: Primary
Antigen: MelanA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR