Tisk…

Anti-MMP1 Rabbit polyclonal antibody

ABCAAB15473-7EA 16480 CZK
ABCAAB15473-7
Anti-MMP1 Rabbit polyclonal antibody
Type: Primary
Antigen: MMP1 (matrix metallopeptidase 1 (interstitial collagenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR