Tisk…

Anti-NSE Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB15486-500EA 27030 CZK
ABCAAB15486-500
Anti-NSE Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NSE
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR