Tisk…

Anti-STAT3 Rabbit polyclonal antibody

ABCAAB15524-7EA 16480 CZK
ABCAAB15524-7
Anti-STAT3 Rabbit polyclonal antibody
Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR